Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAAR1
Entrez Gene ID
708944
Gene Name
trace amine associated receptor 1
Gene Aliases
TAR1
Chromosome Location
Chr.4: 36189646 - 36190662 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080765.1 NP_001074234.1 - 177 152

View Full Product Details