Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFKBIE
Entrez Gene ID
702498
Gene Name
NFKB inhibitor epsilon
Gene Aliases
-
Chromosome Location
Chr.4: 125063907 - 125071778 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002808387.3 XP_002808433.1 - 1218 92
GenBank mRNA JU487310.1 - - 893 92
JV617733.1 - - 1002 92

View Full Product Details