Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPYC
Entrez Gene ID
709645
Gene Name
epiphycan
Gene Aliases
-
Chromosome Location
Chr.11: 90721531 - 90777267 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194215.3 NP_001181144.2 - 886 153

View Full Product Details