Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAS2
Entrez Gene ID
700952
Gene Name
growth arrest specific 2
Gene Aliases
-
Chromosome Location
Chr.14: 42759048 - 42903345 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257624.1 NP_001244553.1 - 417 111
XM_015114619.2 XP_014970105.1 - 804 111
XM_028832580.1 XP_028688413.1 - 810 111
XM_028832581.1 XP_028688414.1 - 807 111
XM_028832582.1 XP_028688415.1 - 806 111
XM_028832583.1 XP_028688416.1 - 804 111
GenBank mRNA JU321049.1 - - 417 111

View Full Product Details