Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACOT8
Entrez Gene ID
717238
Gene Name
acyl-CoA thioesterase 8
Gene Aliases
SNX21
Chromosome Location
Chr.10: 81021687 - 81039558 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266088.1 NP_001253017.1 - 566 99
XM_015148852.2 XP_015004338.1 - 589 99
XM_015148854.2 XP_015004340.1 - 442 99
GenBank mRNA JU327233.1 - - 1684 99
JU472253.1 - - 566 99
JV597667.1 - - 2198 99

View Full Product Details