Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAF1
Entrez Gene ID
700583
Gene Name
TNF receptor associated factor 1
Gene Aliases
EGK_07370
Chromosome Location
Chr.15: 21778553 - 21805403 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265893.1 NP_001252822.1 - 1423 125
XM_015116917.2 XP_014972403.1 - 1687 125
XM_015116918.2 XP_014972404.1 - 1492 125
XM_028834257.1 XP_028690090.1 - 1446 125
GenBank mRNA JU472522.1 - - 1423 125

View Full Product Details