Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NELFA
Entrez Gene ID
712689
Gene Name
negative elongation factor complex member A
Gene Aliases
WHSC2
Chromosome Location
Chr.5: 2044365 - 2072318 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100646.4 XP_001100646.3 - 1892 123
GenBank mRNA JU336258.1 - - 805 123
JV048170.1 - - 852 123
JV627873.1 - - 768 123
JV660585.1 - - 853 123

View Full Product Details