Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS9
Entrez Gene ID
705554
Gene Name
dehydrogenase/reductase 9
Gene Aliases
-
Chromosome Location
Chr.12: 56209504 - 56235852 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028830816.1 XP_028686649.1 - 1090 117

View Full Product Details