Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIB2
Entrez Gene ID
709429
Gene Name
calcium and integrin binding family member 2
Gene Aliases
-
Chromosome Location
Chr.7: 55340645 - 55366202 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257882.1 NP_001244811.1 - 132 87
GenBank mRNA JU332526.1 - - 132 87
JU474000.1 - - 120 87

View Full Product Details