Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPA13
Entrez Gene ID
694660
Gene Name
heat shock protein family A (Hsp70) member 13
Gene Aliases
STCH
Chromosome Location
Chr.3: 32468673 - 32479064 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193708.2 NP_001180637.1 - 665 138
GenBank mRNA JU321543.1 - - 606 138
JU473530.1 - - 593 138
JV044197.1 - - 594 138
JV565952.1 - - 594 138

View Full Product Details