The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001095296.4 | XP_001095296.2 | - | 810 | 65 |
GenBank mRNA | JV044396.1 | - | - | 320 | 65 |
JV663997.1 | - | - | 265 | 65 |