Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPP14
Entrez Gene ID
706788
Gene Name
ribonuclease P/MRP subunit p14
Gene Aliases
EGK_21389
Chromosome Location
Chr.2: 114457021 - 114468914 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266712.1 NP_001253641.1 - 374 145
XM_015130593.2 XP_014986079.1 - 668 145
XM_015130594.2 XP_014986080.1 - 439 145
XM_015130595.2 XP_014986081.1 - 334 145
XM_015130596.2 XP_014986082.1 - 264 145
XM_015130597.2 XP_014986083.1 - 671 145
XM_028843253.1 XP_028699086.1 - 481 145
XM_028843254.1 XP_028699087.1 - 458 145
GenBank mRNA JU332688.1 - - 331 145
JU472057.1 - - 204 145
JU472058.1 - - 548 145
JV044948.1 - - 374 145
JV044949.1 - - 341 145
JV044950.1 - - 374 145

View Full Product Details