Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFAIP6
Entrez Gene ID
694699
Gene Name
TNF alpha induced protein 6
Gene Aliases
EGK_04421
Chromosome Location
Chr.12: 38531363 - 38552242 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265940.1 NP_001252869.1 - 179 121
GenBank mRNA JU322968.1 - - 179 121

View Full Product Details