Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCTN3
Entrez Gene ID
700703
Gene Name
dynactin subunit 3
Gene Aliases
EGK_07599
Chromosome Location
Chr.15: 49583383 - 49590372 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265650.1 NP_001252579.1 - 128 133
XM_015117359.2 XP_014972845.1 - 158 133
XM_015117360.2 XP_014972846.1 - 158 133
GenBank mRNA JV667335.1 - - 128 133

View Full Product Details