Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC33
Entrez Gene ID
696436
Gene Name
tetratricopeptide repeat domain 33
Gene Aliases
-
Chromosome Location
Chr.6: 40567575 - 40611022 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261196.1 NP_001248125.1 - 418 95
GenBank mRNA JU333728.1 - - 412 95
JU474973.1 - - 418 95
JV710937.1 - - 448 95

View Full Product Details