Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLCF1
Entrez Gene ID
712554
Gene Name
cardiotrophin like cytokine factor 1
Gene Aliases
-
Chromosome Location
Chr.14: 7150764 - 7160341 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001106809.4 XP_001106809.1 - 383 120

View Full Product Details