Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HACD1
Entrez Gene ID
700635
Gene Name
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases
PTPLA
Chromosome Location
Chr.9: 18740633 - 18770208 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193912.1 NP_001180841.1 - 814 125
XM_015146546.2 XP_015002032.1 - 837 125
XM_015146547.2 XP_015002033.1 - 734 125

View Full Product Details