Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL36A
Entrez Gene ID
705136
Gene Name
interleukin 36 alpha
Gene Aliases
IL1F6
Chromosome Location
Chr.13: 16186582 - 16189773 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015113172.2 XP_014968658.1 - 682 92

View Full Product Details