Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAML1
Entrez Gene ID
715047
Gene Name
mastermind like transcriptional coactivator 1
Gene Aliases
-
Chromosome Location
Chr.6: 177135558 - 177188638 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001105097.4 XP_001105097.3 - 1759 111
GenBank mRNA JU325855.1 - - 1390 111
JU497055.1 - - 1484 111

View Full Product Details