Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SUPT7L
- Entrez Gene ID
-
702030
- Gene Name
- SPT7 like, STAGA complex subunit gamma
- Gene Aliases
-
EGK_05178
- Chromosome Location
-
Chr.13: 81301869 - 81315452 on Build Mmul_10
- Gene Symbol
- SLC4A1AP
- Entrez Gene ID
-
701783
- Gene Name
- solute carrier family 4 member 1 adaptor protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 81270504 - 81301939 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV645737.1 |
- |
- |
923 |
126 |
View Full Product Details