Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN1
Entrez Gene ID
693541
Gene Name
neuroligin 1
Gene Aliases
EGK_11892
Chromosome Location
Chr.2: 78404863 - 79305883 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257734.1 NP_001244663.1 - 1163 70
XM_015131520.2 XP_014987006.1 - 1834 70
XM_015131521.2 XP_014987007.1 - 1926 70
XM_015131524.2 XP_014987010.1 - 1921 70
XM_015131526.2 XP_014987012.1 - 1502 70
XM_028843069.1 XP_028698902.1 - 1895 70
XM_028843070.1 XP_028698903.1 - 1895 70
XM_028843071.1 XP_028698904.1 - 2567 70
XM_028843072.1 XP_028698905.1 - 2636 70
XM_028843073.1 XP_028698906.1 - 2499 70
XM_028843074.1 XP_028698907.1 - 2499 70
XM_028843075.1 XP_028698908.1 - 2143 70
XM_028843077.1 XP_028698910.1 - 1995 70
GenBank mRNA JU321927.1 - - 1163 70

View Full Product Details