Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDCD3
Entrez Gene ID
699353
Gene Name
NudC domain containing 3
Gene Aliases
EGK_13730
Chromosome Location
Chr.3: 70728249 - 70834677 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266593.1 NP_001253522.1 - 357 120
GenBank mRNA JU333671.1 - - 385 120
JU474924.1 - - 384 120
JV046582.1 - - 357 120
JV710772.1 - - 386 120

View Full Product Details