Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NELFB
Entrez Gene ID
704966
Gene Name
negative elongation factor complex member B
Gene Aliases
COBRA1
Chromosome Location
Chr.15: 930166 - 949913 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015116253.2 XP_014971739.1 - 1790 147
GenBank mRNA JU333872.1 - - 144 147
JU333873.1 - - 144 147
JU472033.1 - - 165 147
JU472034.1 - - 165 147
JV046684.1 - - 295 147
JV046685.1 - - 295 147

View Full Product Details