Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIG1
Entrez Gene ID
700988
Gene Name
androgen induced 1
Gene Aliases
-
Chromosome Location
Chr.4: 46841323 - 47116713 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260565.1 NP_001247494.1 - 439 122
XM_028846704.1 XP_028702537.1 - 579 122
XM_028846705.1 XP_028702538.1 - 637 122
XM_028846706.1 XP_028702539.1 - 473 122
XM_028846707.1 XP_028702540.1 - 553 122
XM_028846709.1 XP_028702542.1 - 215 122
GenBank mRNA JU328131.1 - - 370 122
JV047297.1 - - 439 122
JV601698.1 - - 432 122

View Full Product Details