Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTPAP
Entrez Gene ID
694491
Gene Name
mitochondrial poly(A) polymerase
Gene Aliases
-
Chromosome Location
Chr.9: 31202561 - 31240720 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001083177.4 XP_001083177.1 - 201 134
GenBank mRNA JU323499.1 - - 191 134
JU332288.1 - - 191 134
JU476214.1 - - 165 134

View Full Product Details