Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A10
Entrez Gene ID
696132
Gene Name
solute carrier family 16 member 10
Gene Aliases
-
Chromosome Location
Chr.4: 14732057 - 14870734 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260620.1 NP_001247549.1 - 1401 117
GenBank mRNA JU471273.1 - - 1401 117

View Full Product Details