Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TREM1
Entrez Gene ID
693558
Gene Name
triggering receptor expressed on myeloid cells 1
Gene Aliases
-
Chromosome Location
Chr.4: 128102193 - 128115437 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082517.4 XP_001082517.1 - 698 153

View Full Product Details