Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERTAD4
Entrez Gene ID
712616
Gene Name
SERTA domain containing 4
Gene Aliases
EGK_01691
Chromosome Location
Chr.1: 67723034 - 67736513 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001109829.4 XP_001109829.1 - 573 141
XM_015117531.2 XP_014973017.1 - 410 141

View Full Product Details