Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMSY
Entrez Gene ID
697319
Gene Name
EMSY transcriptional repressor, BRCA2 interacting
Gene Aliases
C14H11orf30, EGK_06698
Chromosome Location
Chr.14: 69507060 - 69611882 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115228.2 XP_014970714.1 - 2747 128
XM_015115229.2 XP_014970715.1 - 2744 128
XM_015115230.2 XP_014970716.1 - 2720 128
XM_015115232.2 XP_014970718.1 - 2717 128
XM_015115233.2 XP_014970719.1 - 2594 128
XM_015115234.2 XP_014970720.1 - 2582 128
XM_015115236.2 XP_014970722.1 - 2567 128
XM_028833834.1 XP_028689667.1 - 2747 128
GenBank mRNA JU335540.1 - - 2698 128
JU476467.1 - - 2698 128
JV719156.1 - - 2703 128

View Full Product Details