Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA10
Entrez Gene ID
718133
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
Gene Aliases
-
Chromosome Location
Chr.14: 64797214 - 64804924 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113644.3 XP_001113644.1 - 2267 77

View Full Product Details