Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH2
Entrez Gene ID
694158
Gene Name
junctophilin 2
Gene Aliases
-
Chromosome Location
Chr.10: 79344374 - 79422766 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001083346.3 XP_001083346.2 - 3091 59
GenBank mRNA JV066346.1 - - 47 59
JV656612.1 - - 278 59

View Full Product Details