Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKEF1
Entrez Gene ID
718476
Gene Name
ankyrin repeat and EF-hand domain containing 1
Gene Aliases
ANKRD5
Chromosome Location
Chr.10: 56491133 - 56513344 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001116097.4 XP_001116097.2 - 2553 140
XM_015149479.2 XP_015004965.1 - 1932 140

View Full Product Details