Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIN7B
Entrez Gene ID
718587
Gene Name
lin-7 homolog B, crumbs cell polarity complex component
Gene Aliases
-
Chromosome Location
Chr.19: 49092534 - 49096597 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194649.1 NP_001181578.1 - 263 130
GenBank mRNA JU320864.1 - - 248 130
JV529545.1 - - 263 130
JV567959.1 - - 272 130
JV668791.1 - - 265 130

View Full Product Details