Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA1
Entrez Gene ID
710462
Gene Name
spermatogenesis associated 1
Gene Aliases
-
Chromosome Location
Chr.1: 139851947 - 139919623 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028831926.1 XP_028687759.1 - 1392 87
XM_028831932.1 XP_028687765.1 - 1392 87
XM_028831937.1 XP_028687770.1 - 1392 87

View Full Product Details