Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAI2
Entrez Gene ID
696977
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
-
Chromosome Location
Chr.16: 70608660 - 70652464 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028836750.1 XP_028692583.1 - 472 109
XM_028836752.1 XP_028692585.1 - 458 109
XM_028836753.1 XP_028692586.1 - 584 109
GenBank mRNA JU491606.1 - - 382 109

View Full Product Details