Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A5
Entrez Gene ID
696967
Gene Name
membrane spanning 4-domains A5
Gene Aliases
EGK_06168
Chromosome Location
Chr.14: 13739928 - 13750505 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086601.4 XP_001086601.1 - 460 127

View Full Product Details