Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC7
Entrez Gene ID
700833
Gene Name
armadillo repeat containing 7
Gene Aliases
-
Chromosome Location
Chr.16: 71455627 - 71481614 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261794.1 NP_001248723.1 - 165 66
XM_015120247.2 XP_014975733.1 - 450 66
XM_015120248.2 XP_014975734.1 - 121 66
XM_015120250.2 XP_014975736.1 - 450 66
GenBank mRNA JU471957.1 - - 165 66

View Full Product Details