Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC7
Entrez Gene ID
703164
Gene Name
coiled-coil domain containing 7
Gene Aliases
C9H10orf68
Chromosome Location
Chr.9: 33382314 - 33823161 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028826154.1 XP_028681987.1 - 4577 140

View Full Product Details