Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASRGL1
Entrez Gene ID
718871
Gene Name
asparaginase and isoaspartyl peptidase 1
Gene Aliases
EGK_06117
Chromosome Location
Chr.14: 11810555 - 11843927 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194716.3 NP_001181645.1 - 444 87
GenBank mRNA JU323185.1 - - 465 87
JU323186.1 - - 465 87
JV045810.1 - - 461 87

View Full Product Details