Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTI2
Entrez Gene ID
696776
Gene Name
TELO2 interacting protein 2
Gene Aliases
C8orf41
Chromosome Location
Chr.8: 33844213 - 33872432 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193802.3 NP_001180731.2 - 1257 114
XM_015145053.2 XP_015000539.1 - 1240 114
GenBank mRNA JU336483.1 - - 897 114
JU472787.1 - - 996 114
JV044754.1 - - 1020 114

View Full Product Details