Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PROSER1
Entrez Gene ID
696606
Gene Name
proline and serine rich 1
Gene Aliases
-
Chromosome Location
Chr.17: 17733964 - 17763418 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087439.4 XP_001087439.2 - 1413 102
XM_015120959.2 XP_014976445.2 - 745 102
GenBank mRNA JU477301.1 - - 1401 102
JU477302.1 - - 1401 102
JV048305.1 - - 738 102
JV724005.1 - - 1063 102

View Full Product Details