Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A21
Entrez Gene ID
697370
Gene Name
solute carrier family 25 member 21
Gene Aliases
EGK_18128
Chromosome Location
Chr.7: 98746108 - 99257818 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265644.1 NP_001252573.1 - 1130 150
GenBank mRNA JV636440.1 - - 1130 150

View Full Product Details