Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSDMC
Entrez Gene ID
695343
Gene Name
gasdermin C
Gene Aliases
EGK_19278
Chromosome Location
Chr.8: 130068986 - 130097939 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028852908.1 XP_028708741.1 - 404 120

View Full Product Details