Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOK6
Entrez Gene ID
703667
Gene Name
docking protein 6
Gene Aliases
-
Chromosome Location
Chr.18: 10089307 - 10523752 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266038.1 NP_001252967.1 - 739 72
XM_028837739.1 XP_028693572.1 - 786 72
GenBank mRNA JU320219.1 - - 739 72
JV574491.1 - - 778 72

View Full Product Details