Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELMOD2
Entrez Gene ID
698742
Gene Name
ELMO domain containing 2
Gene Aliases
EGK_16116
Chromosome Location
Chr.5: 138401156 - 138430860 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193877.2 NP_001180806.1 - 654 107
XM_015139222.2 XP_014994708.1 - 674 107
GenBank mRNA JU483866.1 - - 648 107

View Full Product Details