Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP11
Entrez Gene ID
698835
Gene Name
aquaporin 11
Gene Aliases
-
Chromosome Location
Chr.14: 70666390 - 70684651 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266495.2 NP_001253424.1 - 883 136
GenBank mRNA JV046799.1 - - 884 136
JV578877.1 - - 922 136

View Full Product Details