Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP4F22
Entrez Gene ID
718303
Gene Name
cytochrome P450, family 4, subfamily F, polypeptide 22
Gene Aliases
-
Chromosome Location
Chr.19: 15106415 - 15149547 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194539.2 NP_001181468.2 - 423 63

View Full Product Details