Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL10
Entrez Gene ID
696524
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
-
Chromosome Location
Chr.6: 173811651 - 173825836 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266393.1 NP_001253322.1 - 623 149
GenBank mRNA JU321021.1 - - 623 149

View Full Product Details