Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCP2
Entrez Gene ID
704228
Gene Name
Purkinje cell protein 2
Gene Aliases
EGK_10035
Chromosome Location
Chr.19: 7573468 - 7578891 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127693.1 NP_001121165.1 - 52 99
XM_015122684.2 XP_014978170.1 - 1072 99
GenBank mRNA EU676240.1 - - 52 99
JV044561.1 - - 267 99
JV642073.1 - - 273 99

View Full Product Details