Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMC10
Entrez Gene ID
719285
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
C19H19orf63, C19orf63
Chromosome Location
Chr.19: 50383940 - 50391420 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194699.2 NP_001181628.1 - 329 141
XM_015124415.2 XP_014979901.1 - 394 141
GenBank mRNA JU322500.1 - - 375 141
JU484832.1 - - 460 141
JV045277.1 - - 369 141
JV579089.1 - - 378 141

View Full Product Details